Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-10
Supplier:  Biolegend 504912
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-10 [JES5-2A5]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT Capture, Neut, WB; Size: 100 mg
Catalog No. 50-208-5662