Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-12/IL-23 p40
Supplier:  Biolegend 505309
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-12/IL-23 p40 [C17.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA, Neut, ICFC, IP, WB; Size: 5 mg
Catalog No. 50-208-5678