Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-2
Supplier:  Biolegend 503709
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-2 [JES6-1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT Capture, IP, Neut, WB; Size: 50 mg
Catalog No. 50-208-5612