Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-23 (p19)
Supplier:  Biolegend 513808
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-23 (p19) [MMp19B2]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: Neut, ELISA Capture, ICC; Size: 1 mg
Catalog No. 50-208-5865