Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse/rat CD61
Supplier:  Biolegend 104325
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse/rat CD61 [2C9.G2 (HMβ3-1)]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, Block, Activ, IHC; Size: 100 ug
Catalog No. 50-206-9811