Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse TCR β chain
Supplier:  Biolegend 109255
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF, IHC-F, IP, Costim, Depletion; Size: 5 mg
Catalog No. 50-207-0087