Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116253
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, IHC-P, IP, WB, CMCD; Size: 100 ug
Catalog No. 50-207-0264