Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse Tim-4
Supplier:  Biolegend 130013
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse Tim-4 [RMT4-54]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, FA, Block; Size: 100 ug
Catalog No. 50-207-0903