Learn More
Biolegend Ultra-LEAF™ Purified anti-rat CD90
Supplier:  Biolegend 206204
Encompass_Preferred
Ultra-LEAF™ Purified anti-rat CD90 [13H7]; Isotype: Mouse IgG3, κ; Reactivity: Rat; Apps: FC, IP, WB; Size: 500 ug
Catalog No. 50-207-2543