Learn More
Biolegend Ultra-LEAF™ Purified anti-SARS-CoV-2 S Protein S1 [A20103J]; Isotype: Rat IgG2a, κ; Reactivity: SARS-CoV-2; Apps: Block; Size: 1 mg
Supplier:  Biolegend 944704
Encompass_Preferred
Ultra-LEAF™ Purified anti-SARS-CoV-2 S Protein S1 [A20103J]; Isotype: Rat IgG2a, κ; Reactivity: SARS-CoV-2; Apps: Block; Size: 1 mg
Catalog No. 50-237-7154