Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgG2a, κ isotype Ctrl
Supplier:  Biolegend 400281
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 5 mg
Catalog No. NC1422782