Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgG1, κ Isotype Ctrl
Supplier:  Biolegend 401408
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgG1, κ Isotype Ctrl [MG1-45]; Isotype: Mouse (BALB/c) IgG1, κ; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 1 mg
Catalog No. 50-403-578