Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgG2b, κ Isotype Ctrl
Supplier:  Biolegend 401215
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgG2b, κ Isotype Ctrl [MG2b-57]; Isotype: Mouse IgG2b, κ; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 100 ug
Catalog No. 50-207-9596