Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgG3, κ isotype Ctrl
Supplier:  Biolegend 401327
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgG3, κ isotype Ctrl [MG3-35]; Isotype: Mouse IgG3, κ; Apps: FC; Size: 100 ug
Catalog No. 50-207-9604