Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgM, κ Isotype Ctrl
Supplier:  Biolegend 401633
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgM, κ Isotype Ctrl [MM-30]; Isotype: Mouse IgM, κ; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 50 mg
Catalog No. 50-207-9631