Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-6 Recombinant
Supplier:  Biolegend 941404
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-6 Recombinant [QA19A65]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: Neut; Size: 1 mg
Catalog No. 50-207-5433