Learn More
Biolegend Ultra-LEAF™ Purified Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400574
Encompass_Preferred
Ultra-LEAF™ Purified Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 100 mg
Catalog No. 50-207-9519