Learn More
Biolegend Ultra-LEAF™ Purified Rat IgG1, κ isotype Ctrl
Supplier:  Biolegend 400431
Encompass_Preferred
Ultra-LEAF™ Purified Rat IgG1, κ isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Apps: FC, ICFC, ICC, IHC, WB, IP, FA; Size: 100 ug
Catalog No. 50-207-9481