Learn More
Biolegend Ultra-LEAF™ Purified Rat IgG2b, κ Isotype Ctrl
Supplier:  Biolegend 400643
Encompass_Preferred
Ultra-LEAF™ Purified Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 100 ug
Catalog No. 50-207-9533