Learn More
Biolegend Ultra-LEAF™ Purified Syrian Hamster IgG Isotype Ctrl
Supplier:  Biolegend 402025
Encompass_Preferred
Ultra-LEAF™ Purified Syrian Hamster IgG Isotype Ctrl [SHG-1]; Isotype: Syrian Hamster IgG; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 5 mg
Catalog No. 50-207-9652