Learn More
Bioworld Ulex europaeus (Gorse) Lectin (UEA I) - FITC
Catalog No. 50199173
Encompass
Technical Specifications:Ligand Source:Ulex europaeusMolecular Weight: 63 kDaCarbohydrate Specificity:Fucα1,3Galβ4GlcNAc (H-type 2)Inhibiting/Eluting Sugar: FucoseCarbohydrate for Elution: 0.2M FucoseBlood Group Specificity: O>A2Conjugate: FITCForm: LiquidIsoelectric (pI): 4.5-5.1Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.
Catalog No. 50-199-173 Supplier Bioworld Supplier No. 217610621
Technical Specifications:Ligand Source:Ulex europaeusMolecular Weight: 63 kDaCarbohydrate Specificity:Fucα1,3Galβ4GlcNAc (H-type 2)Inhibiting/Eluting Sugar: FucoseCarbohydrate for Elution: 0.2M FucoseBlood Group Specificity: O>A2Conjugate: FITCForm: LiquidIsoelectric (pI): 4.5-5.1Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.