Learn More
Bioworld Ulex europaeus (Gorse) Lectin (UEA I) - Rhodamine (TRITC)
Catalog No. 50199174
Encompass
Technical Specifications:Ligand Source:Ulex europaeusMolecular Weight:63 kDaCarbohydrate Specificity:Fucα1,3Galβ4GlcNAc (H-type 2)Inhibiting/Eluting Sugar: FucoseCarbohydrate for Elution:0.2M FucoseBlood Group Specificity:O>A2Conjugate: RhodamineForm:LiquidIsoelectric (pI):4.5-5.1Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.
Catalog No. 50-199-174 Supplier Bioworld Supplier No. 217610631
Technical Specifications:Ligand Source:Ulex europaeusMolecular Weight:63 kDaCarbohydrate Specificity:Fucα1,3Galβ4GlcNAc (H-type 2)Inhibiting/Eluting Sugar: FucoseCarbohydrate for Elution:0.2M FucoseBlood Group Specificity:O>A2Conjugate: RhodamineForm:LiquidIsoelectric (pI):4.5-5.1Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.