Learn More
Uline 2 x 3"" 6 Mil Reclosable Bags, 1000 pack
Catalog No. NC1626777
Encompass_Preferred
Superior strength, puncture-resistant poly bags with heavy-duty zipper for security. Recommended for auto and machinery parts, electrical panels, printing plates and tools. Archival quality, acid free for long-term storage. 100% virgin polyethylene film. Meets FDA and USDA specifications.
Catalog No. NC1626777 Supplier Uline Supplier No. S12293
Superior strength, puncture-resistant poly bags with heavy-duty zipper for security. Recommended for auto and machinery parts, electrical panels, printing plates and tools. Archival quality, acid free for long-term storage. 100% virgin polyethylene film. Meets FDA and USDA specifications.