Learn More
Medchemexpress LLC Ulipristal | 159811-51-5 | ≥98.0% | 433.58 | 100 MG
SDP
Catalog No. 5000360145
Encompass_Preferred
Ulipristal is a selective progesterone receptor modulator (SPRM). It functions by binding to the progesterone receptor, which inhibits PR-mediated gene expression and interferes with progesterone activity in the reproductive system.
  • May suppress the growth of uterine leiomyomatosis.
  • Can be used as an emergency contraception.
  • Inhibits or delays ovulation.
  • Affects endometrial tissue.
Catalog No. 50-003-60145 Supplier Medchemexpress LLC Supplier No. HY14959100MG
Ulipristal is a selective progesterone receptor modulator (SPRM). It functions by binding to the progesterone receptor, which inhibits PR-mediated gene expression and interferes with progesterone activity in the reproductive system.
  • May suppress the growth of uterine leiomyomatosis.
  • Can be used as an emergency contraception.
  • Inhibits or delays ovulation.
  • Affects endometrial tissue.