Learn More
Medchemexpress LLC Ulipristal | 159811-51-5 | ≥98.0% | 433.58 | 50 MG
SDP
Catalog No. 5000361858
Encompass_Preferred
Ulipristal is a selective progesterone receptor modulator (SPRM) that binds to the progesterone receptor, inhibiting PR-mediated gene expression and interfering with progesterone activity in the reproductive system.
  • Selective progesterone receptor modulator (SPRM)
  • Binds to the progesterone receptor
  • Inhibits PR-mediated gene expression
  • Interferes with progesterone activity in the reproductive system
  • May suppress the growth of uterine leiomyomatosis
  • Can be used as emergency contraception by inhibiting or delaying ovulation and affecting endometrial tissue
Catalog No. 50-003-61858 Supplier Medchemexpress LLC Supplier No. HY1495950MG
Ulipristal is a selective progesterone receptor modulator (SPRM) that binds to the progesterone receptor, inhibiting PR-mediated gene expression and interfering with progesterone activity in the reproductive system.
  • Selective progesterone receptor modulator (SPRM)
  • Binds to the progesterone receptor
  • Inhibits PR-mediated gene expression
  • Interferes with progesterone activity in the reproductive system
  • May suppress the growth of uterine leiomyomatosis
  • Can be used as emergency contraception by inhibiting or delaying ovulation and affecting endometrial tissue