Learn More
Medchemexpress LLC Ulipristal acetate | 126784-99-4 | 475.62 | 100 MG
SDP
Catalog No. 5000394088
Encompass_Preferred
Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). It stimulates the autophagic response selectively in leiomyoma cells and has potential for treating benign gynecological conditions like uterine myoma.
  • Orally active, selective progesterone receptor modulator
  • Stimulates autophagy in leiomyoma cells
  • Potential for treating benign gynecological conditions, such as uterine myoma
  • Blocks activin A modulation of fibronectin and vascular endothelial growth factor A (VEGF-A) mRNA expression
  • Possesses significant antiprogestational activity in vivo
  • Decreases incidences of fibroadenomas and adenocarcinomas in the mammary gland
Catalog No. 50-003-94088 Supplier Medchemexpress LLC Supplier No. HY16508100MG
Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). It stimulates the autophagic response selectively in leiomyoma cells and has potential for treating benign gynecological conditions like uterine myoma.
  • Orally active, selective progesterone receptor modulator
  • Stimulates autophagy in leiomyoma cells
  • Potential for treating benign gynecological conditions, such as uterine myoma
  • Blocks activin A modulation of fibronectin and vascular endothelial growth factor A (VEGF-A) mRNA expression
  • Possesses significant antiprogestational activity in vivo
  • Decreases incidences of fibroadenomas and adenocarcinomas in the mammary gland