Learn More
Apexbio Technology LLC Ulipristal acetate 126784-99-4 50mg
SDP
Catalog No. 5000092566
Encompass_Preferred
Ulipristal acetate (126784-99-4) is a selective progesterone receptor modulator (SPRM) targeting progesterone receptors It is designed to modulate receptor-mediated signaling thereby regulating endometrial tissue proliferation and fibroid cell growth Ulipristal acetate exerts its biological activity primarily through reversible inhibition of progesterone-dependent signaling pathways Based on these pharmacological properties ulipristal acetate holds research potential in studies of progesterone signaling pathways uterine physiology hormonal regulation mechanisms and benign gynecological conditions such as uterine myoma
Catalog No. 50-000-92566 Supplier Apexbio Technology LLC Supplier No. B104950
Ulipristal acetate (126784-99-4) is a selective progesterone receptor modulator (SPRM) targeting progesterone receptors It is designed to modulate receptor-mediated signaling thereby regulating endometrial tissue proliferation and fibroid cell growth Ulipristal acetate exerts its biological activity primarily through reversible inhibition of progesterone-dependent signaling pathways Based on these pharmacological properties ulipristal acetate holds research potential in studies of progesterone signaling pathways uterine physiology hormonal regulation mechanisms and benign gynecological conditions such as uterine myoma