Learn More
TARGETMOL CHEMICALS INC ULIPRISTAL ACETATE 50MG
SDP
Catalog No. 502577098
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ulipristal acetate (CDB-2914) is an orally bioavailable acetate salt of ulipristal a selective progesterone receptor modulator with anti-progesterone activity. Ulipristal binds to the progesterone receptor (PR) thereby inhibiting PR-mediated gene expression and interfering with progesterone activity in the reproductive system. As a result this agent may suppress the growth of uterine leiomyomatosis. Furthermore by inhibiting or delaying ovulation and effecting endometrial tissue ulipristal can be used as an emergency contraception. purity: 100%
Catalog No. 50-257-7098 Supplier TARGETMOL CHEMICALS INC Supplier No. T252750MG
Also available in 5 mg 10 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ulipristal acetate (CDB-2914) is an orally bioavailable acetate salt of ulipristal a selective progesterone receptor modulator with anti-progesterone activity. Ulipristal binds to the progesterone receptor (PR) thereby inhibiting PR-mediated gene expression and interfering with progesterone activity in the reproductive system. As a result this agent may suppress the growth of uterine leiomyomatosis. Furthermore by inhibiting or delaying ovulation and effecting endometrial tissue ulipristal can be used as an emergency contraception. purity: 100%