Learn More
Medchemexpress LLC HY-16508 5mg Medchemexpress, Ulipristal acetate CAS:126784-99-4 Purity:>98%
SDP
Catalog No. 502030719
Encompass_Preferred
Medchemexpress, HY-16508 5mg Ulipristal acetate CAS:126784-99-4 Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). Ulipristal acetate stimulates the autophagic response selectively in leiomyoma cells. Ulipristal acetate has the potential for benign gynecological conditions treatment, such as uterine myoma[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0719 Supplier Medchemexpress LLC Supplier No. HY165085MG
Medchemexpress, HY-16508 5mg Ulipristal acetate CAS:126784-99-4 Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). Ulipristal acetate stimulates the autophagic response selectively in leiomyoma cells. Ulipristal acetate has the potential for benign gynecological conditions treatment, such as uterine myoma[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.