Learn More
Medchemexpress LLC Ulipristal acetate (CDB-2914) | 126784-99-4 | 99.9% | 475.62 | 1 ML
SDP
Catalog No. 5000394133
Encompass_Preferred
Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). It stimulates the autophagic response selectively in leiomyoma cells and has potential for treating benign gynecological conditions like uterine myoma.
  • Orally active.
  • Selective progesterone receptor modulator (SPRM).
  • Stimulates autophagic response in leiomyoma cells.
  • Potential for treating uterine myoma.
Catalog No. 50-003-94133 Supplier Medchemexpress LLC Supplier No. HY1650810MM/1ML
Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). It stimulates the autophagic response selectively in leiomyoma cells and has potential for treating benign gynecological conditions like uterine myoma.
  • Orally active.
  • Selective progesterone receptor modulator (SPRM).
  • Stimulates autophagic response in leiomyoma cells.
  • Potential for treating uterine myoma.