Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ulixacaltamide-d9 hydrochloride | 1346604-21-4 | 98.0% | 1 MG
SDP

Catalog No. 5000234536
Encompass_Preferred

Ulixacaltamide-d9 hydrochloride is the deuterium-labeled hydrochloride salt of ulixacaltamide, supplied for research use as an isotopically labeled reference or tracer in biochemical and pharmacological studies.

  • Deuterium-labeled tracer for analytical and pharmacokinetic studies
  • Hydrochloride salt with deuterium incorporation for isotopic studies
  • Reported molecular weight 429.40 g/mol and formula C19H19D9Cl2FN3O2
  • Provided in small research pack sizes suitable for assay development and reference standards
  • For research use only; not for human use

Catalog No. 50-002-34536 Supplier Medchemexpress LLC Supplier No. HY120546S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ulixacaltamide-d9 hydrochloride is the deuterium-labeled hydrochloride salt of ulixacaltamide, supplied for research use as an isotopically labeled reference or tracer in biochemical and pharmacological studies.

  • Deuterium-labeled tracer for analytical and pharmacokinetic studies
  • Hydrochloride salt with deuterium incorporation for isotopic studies
  • Reported molecular weight 429.40 g/mol and formula C19H19D9Cl2FN3O2
  • Provided in small research pack sizes suitable for assay development and reference standards
  • For research use only; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.