Learn More
Chemie Tek ULIXERTINIB HCL, 869886-67-9, MFCD22628898, 10 mg
SDP
Supplier:  Chemie Tek CTVRT75210MG
Encompass_Preferred
Formula: C21H22Cl2N4O2 ∙ HCl, MW: 469.8, Synonym: BVD-523, VRT752271. Ulixertinib (BVD-523, VRT752271) is a cell permeable small-molecule ERK1/2 kinase inhibitor currently under clinical investigation (Phase 1).
Catalog No. NC1259278