Learn More
Medchemexpress LLC Ulixertinib hydrochloride | 1956366-10-1 | MFCD22543853 | 99.9% | 469.79 g/mol | C21H23Cl3N4O2 | 10 MG
SDP
Catalog No. 5000288030
Encompass_Preferred
Ulixertinib hydrochloride is a potent, selective, ATP-competitive ERK1/2 inhibitor provided as a high-purity research chemical for in vitro and cellular pathway studies. The compound is supplied as a solid and requires appropriate storage and solvent handling for stability and solubility.
  • Potent and selective ERK1/2 inhibition (IC50 <0.3 nM against ERK2)
  • High purity (99.89%) suitable for analytical and biological assays
  • White to off-white solid form for accurate weighing and formulation
  • High solubility in DMSO (100 mg/mL); insoluble in water
  • Stable when stored sealed at 4°C; in solvent: -80°C for 2 years, -20°C for 1 year
  • Intended for research use only; not for human or clinical use
Catalog No. 50-002-88030 Supplier Medchemexpress LLC Supplier No. HY15816A10MG
Ulixertinib hydrochloride is a potent, selective, ATP-competitive ERK1/2 inhibitor provided as a high-purity research chemical for in vitro and cellular pathway studies. The compound is supplied as a solid and requires appropriate storage and solvent handling for stability and solubility.
  • Potent and selective ERK1/2 inhibition (IC50 <0.3 nM against ERK2)
  • High purity (99.89%) suitable for analytical and biological assays
  • White to off-white solid form for accurate weighing and formulation
  • High solubility in DMSO (100 mg/mL); insoluble in water
  • Stable when stored sealed at 4°C; in solvent: -80°C for 2 years, -20°C for 1 year
  • Intended for research use only; not for human or clinical use