Learn More
Medchemexpress LLC Ulixertinib (Standard) | 869886-67-9 | 99.9% | 433.33 | 200 MG
SDP
Catalog No. 5000365866
Encompass_Preferred
Ulixertinib is a potent, orally active, and highly selective ATP-competitive, reversible covalent inhibitor of ERK1/2 kinases. It effectively inhibits ERK2 with an IC50 of <0.3 nM, reducing phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) activity in A375 melanoma cells.
  • Potent, orally active, and highly selective
  • ATP-competitive and reversible covalent inhibitor of ERK1/2 kinases
  • Inhibits ERK2 with an IC50 of <0.3 nM
  • Reduces phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) activity
  • Demonstrates antiproliferative activity in various cell lines
  • Investigated in clinical trials for multiple conditions, including solid tumors and various cancers
Catalog No. 50-003-65866 Supplier Medchemexpress LLC Supplier No. 200MGHY15816
Ulixertinib is a potent, orally active, and highly selective ATP-competitive, reversible covalent inhibitor of ERK1/2 kinases. It effectively inhibits ERK2 with an IC50 of <0.3 nM, reducing phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) activity in A375 melanoma cells.
  • Potent, orally active, and highly selective
  • ATP-competitive and reversible covalent inhibitor of ERK1/2 kinases
  • Inhibits ERK2 with an IC50 of <0.3 nM
  • Reduces phosphorylated ERK2 (pERK) and downstream kinase RSK (pRSK) activity
  • Demonstrates antiproliferative activity in various cell lines
  • Investigated in clinical trials for multiple conditions, including solid tumors and various cancers