Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ULK-101 100mg | 2443816-45-1 | 100 MG
SDP

Catalog No. 5000228200
Encompass_Preferred

ULK-101 is a potent and selective ULK1 inhibitor used in research to suppress autophagy and sensitize cancer cells to nutrient stress. It demonstrates high potency against ULK1 with reduced activity against ULK2.

  • Selective ULK1 inhibition with IC50 ≈ 1.6 nM.
  • Reduced activity against ULK2 (IC50 ≈ 30 nM).
  • High chemical purity reported at 99.91%.
  • Molecular formula C22H16F4N4OS and molecular weight 460.45.
  • Supplied as a powder; common pack size 100 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-28200 Supplier Medchemexpress LLC Supplier No. HY114490100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ULK-101 is a potent and selective ULK1 inhibitor used in research to suppress autophagy and sensitize cancer cells to nutrient stress. It demonstrates high potency against ULK1 with reduced activity against ULK2.

  • Selective ULK1 inhibition with IC50 ≈ 1.6 nM.
  • Reduced activity against ULK2 (IC50 ≈ 30 nM).
  • High chemical purity reported at 99.91%.
  • Molecular formula C22H16F4N4OS and molecular weight 460.45.
  • Supplied as a powder; common pack size 100 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.