Learn More
Medchemexpress LLC ULK-101 1mg | 2443816-45-1 | 1 MG
SDP
Catalog No. 5000230074
Encompass_Preferred
ULK-101 is a potent, selective small-molecule inhibitor of ULK1 (unc-51-like autophagy activating kinase 1) that suppresses autophagy and can sensitize cancer cells to nutrient stress. It is supplied as a research-grade solid for in vitro studies.
  • Potent ULK1 inhibition; reported low-nanomolar IC50 for ULK1, ULK2 ~30 nM
  • Suppresses autophagy and sensitizes cancer cells to nutrient stress
  • Molecular formula C22H16F4N4OS and molecular weight ~460.4 g/mol
  • Provided as a 1 mg quantity suitable for assay development and biochemical studies
Catalog No. 50-002-30074 Supplier Medchemexpress LLC Supplier No. HY1144901MG
ULK-101 is a potent, selective small-molecule inhibitor of ULK1 (unc-51-like autophagy activating kinase 1) that suppresses autophagy and can sensitize cancer cells to nutrient stress. It is supplied as a research-grade solid for in vitro studies.
  • Potent ULK1 inhibition; reported low-nanomolar IC50 for ULK1, ULK2 ~30 nM
  • Suppresses autophagy and sensitizes cancer cells to nutrient stress
  • Molecular formula C22H16F4N4OS and molecular weight ~460.4 g/mol
  • Provided as a 1 mg quantity suitable for assay development and biochemical studies