Learn More
Selleck Chemical LLC ULK-101 2mg
SDP
Supplier:  Selleck Chemical LLC S87932MG
Encompass_Preferred
ULK-101 is a potent and selective ULK1 inhibitor with IC50 values of 8.3 nM and 30 nM for ULK1 and ULK2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8200