Learn More
Sino Biological Recombinant Human TP53RK Protein (Full Length, GST Tag)
Catalog No. 502572743
Encompass_Preferred
Recombinant full length human TP53RK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2743 Supplier Sino Biological Supplier No. T2934G100
Recombinant full length human TP53RK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.