Learn More
Selleck Chemical LLC SBI-0206965 5mg 1884220-36-3
SDP
Catalog No. NC1172994
Encompass_Preferred
SBI-0206965 is a highly selective autophagy kinase ULK1 inhibitor with IC50 of 108 nM, about 7-fold selectivity over ULK2. SBI-0206965 inhibits autophagy and enhances apoptosis in human glioblastoma and lung cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1172994 Supplier Selleck Chemical LLC Supplier No. S78855MG
SBI-0206965 is a highly selective autophagy kinase ULK1 inhibitor with IC50 of 108 nM, about 7-fold selectivity over ULK2. SBI-0206965 inhibits autophagy and enhances apoptosis in human glioblastoma and lung cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.