Learn More
Sino Biological Recombinant Human TRIB1 Protein (Full Length, GST Tag)
Catalog No. 502572736
Encompass_Preferred
Recombinant full length human TRIB1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2736 Supplier Sino Biological Supplier No. T3030G1000
Recombinant full length human TRIB1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.