Learn More
Carnabio Usa Inc ULK3/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 051735
Encompass
ULK3, Full-length, Wild type, Amino Acid 1-472(end), NP_001092906.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0656