Learn More
Medchemexpress LLC Ulotaront hydrochloride | 1310422-41-3 | 99.9% | 219.73 | 10 MG
SDP
Catalog No. 5000365331
Encompass_Preferred
Ulotaront hydrochloride is an orally active TAAR1 and 5-HT1A agonist and a CNS active psychotropic agent. It possesses a unique mechanism of action that does not involve D2/5-HT2A receptors, exhibiting antipsychotic-like effects. This compound has potential for the study of schizophrenia.
  • Orally active TAAR1 and 5-HT1A agonist
  • CNS active psychotropic agent
  • Unique mechanism of action, non-D2/5-HT2A
  • Exerts antipsychotic-like effects
  • Potential for the study of schizophrenia
Catalog No. 50-003-65331 Supplier Medchemexpress LLC Supplier No. 10MGHY136109
Ulotaront hydrochloride is an orally active TAAR1 and 5-HT1A agonist and a CNS active psychotropic agent. It possesses a unique mechanism of action that does not involve D2/5-HT2A receptors, exhibiting antipsychotic-like effects. This compound has potential for the study of schizophrenia.
  • Orally active TAAR1 and 5-HT1A agonist
  • CNS active psychotropic agent
  • Unique mechanism of action, non-D2/5-HT2A
  • Exerts antipsychotic-like effects
  • Potential for the study of schizophrenia