Learn More
Tiger Vac ULPA WET/DRY VAC EXP1-10 CR
Catalog No. NC2209284
Encompass
ULPA wet/dry vac, explosion proof, and can work with flammable liquids. The EXP1-10 CR (MRAC)
This item is not returnable. View return policy
Catalog No. NC2209284 Supplier Tiger Vac Supplier No. 110343A
Add to Cart
Add to Cart
Request a Quote
ULPA wet/dry vac, explosion proof, and can work with flammable liquids. The EXP1-10 CR (MRAC)