Learn More
Hach Company Stablcal ULR Verification Kit, 100mL
Supplier:  Hach Company 2714600
Encompass
Stablcal Verification Kit includes: 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 mL each), sililcone oil and oiling cloth, and instructions. For verification only. Not for calibration.
Catalog No. 50-199-6592