Learn More
Beckman Coulter ULT.-CENT. TUBES-NO CAP 25/BX
Catalog No. NC9911362
Encompass_Preferred
Thickwall Polycarbonate Tubes,Quantity:25,Size:38 x 102, Vol.(mL): 94.0
Catalog No. NC9911362 Supplier Beckman Coulter Supplier No. 355628
Thickwall Polycarbonate Tubes,Quantity:25,Size:38 x 102, Vol.(mL): 94.0