Learn More
PHCbi PHCbi MDF-DU702VHA-PA, ENERGY STAR Certified, Ultra-Low Temperature Freezer, 25.7 cu.ft.
Catalog No. NC3718737
Encompass_Preferred
The PHCbi MDF-DU702VHA-PA is an ENERGY STAR certified ultra-low temperature freezer that effectively preserves critical biological samples. With a capacity of 25.7 cu.ft. and a temperature range of -50°C to -86°C, it combines exceptional energy efficiency with advanced insulation technology. Its user-friendly full-color LCD touchscreen enhances monitoring and data management, making it a superior choice for laboratories focused on sustainability and reliability.
Catalog No. NC3718737 Supplier PHCbi Supplier No. MDFDU702VHAPA SCRI
Add to Cart
Add to Cart
Request a Quote
The PHCbi MDF-DU702VHA-PA is an ENERGY STAR certified ultra-low temperature freezer that effectively preserves critical biological samples. With a capacity of 25.7 cu.ft. and a temperature range of -50°C to -86°C, it combines exceptional energy efficiency with advanced insulation technology. Its user-friendly full-color LCD touchscreen enhances monitoring and data management, making it a superior choice for laboratories focused on sustainability and reliability.