Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD40
Supplier:  Biolegend 313019
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD40 [HB14]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, Costim; Size: 100 ug
Catalog No. NC1744883