Learn More
Sciencell Research Laboratories ULT-LOW BIND CULTR PLT,48WELL
Catalog No. NC1866773
Encompass
Ultra-Low Binding Culture Plate (48-well)Includes one Ultra-Low Binding Culture Plate and uses a 48-well plate. Each plate comes individually wrapped and sterile.The Ultra-Low Binding Culture Plates prevent cell attachment by providing a neutrally charged, hydrophilic surface. Each plate is specially treated with a hydrogel that is animal-component free, inert and biocompatible. These plates are useful for the cultures of human mammary stem cells, mammospheres, progenitor cells, neural stem cells, and neurospheres. Additionally, these plates support the efficient formation of three-dimensional (3D) spheroids, such as embryoid body, tumorspheres in cancer research, and the 3D primary spheroids in regenerative medicines.
Catalog No. NC1866773 Supplier Sciencell Research Laboratories Supplier No. 0343
Ultra-Low Binding Culture Plate (48-well)Includes one Ultra-Low Binding Culture Plate and uses a 48-well plate. Each plate comes individually wrapped and sterile.The Ultra-Low Binding Culture Plates prevent cell attachment by providing a neutrally charged, hydrophilic surface. Each plate is specially treated with a hydrogel that is animal-component free, inert and biocompatible. These plates are useful for the cultures of human mammary stem cells, mammospheres, progenitor cells, neural stem cells, and neurospheres. Additionally, these plates support the efficient formation of three-dimensional (3D) spheroids, such as embryoid body, tumorspheres in cancer research, and the 3D primary spheroids in regenerative medicines.