Learn More
Azzota Corp Ultra-Micro Uv-Transparent Disposable Cuvettes, Dimensions 12.5X12.5mm, 15mm Height, 10mm Lightpath, Filling Volume 70L To 550L, Window Dimensions 2X3.5mm, 100Pk
Catalog No. 501612758
Encompass_Preferred
Ultra-micro UV-Transparent Disposable Cuvettes, Dimensions 12.5x12.5mm, 15mm height, 10mm Lightpath, Filling Volume 70L to 550L, Window Dimensions 2x3.5mm, 100pk
Catalog No. 50-161-2758 Supplier Azzota Corp Supplier No. A905
Ultra-micro UV-Transparent Disposable Cuvettes, Dimensions 12.5x12.5mm, 15mm height, 10mm Lightpath, Filling Volume 70L to 550L, Window Dimensions 2x3.5mm, 100pk