Learn More
Crystal Chem Inc Ultra Sensitive Mouse Insulin ELISA Kit (10 Kit Pack), 96-wells per kit, individually packaged
SDP
Supplier:  Crystal Chem Inc 90082
Encompass
The 10-kit pack is an economical option for high-volume users of our Ultra Sensitive Mouse Insulin ELISA. Each kit is idividually packaged for ease of use. Highlights: Kits use only 5 µL sample. Dynamic range of 0.1- 64 ng/mL. High sensitivity of 50 pg/mL when using 5µL sample. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7921